Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC10 Mouse anti-Human, Clone: OTI3C2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525013

Catalog No. PIMA525013


Add to cart

Description

Description

A novel class II histone deacetylase, HDAC10 is most similar to HDAC6. Both contain a unique, putative second catalytic domain not found in other HDACs. In HDAC10, however, this domain is not functional. Unlike HDAC6, which is normally a cytoplasmic deacetylase, HDAC10 resides in both the nucleus and cytoplasm. Data suggests that HDAC10 may play roles both in the nucleus, as a transcriptional modulator, and in the cytoplasm in an unidentified role. HDAC10 variants seem present in certain cell lines and tissues that are as of yet not characterized.
Specifications

Specifications

HDAC10
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q969S8
RP3-402G11.7, DKFZp761B039, MGC149722
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83933
Flow Cytometry, Western Blot
OTI3C2
Unconjugated
HDAC10
Liquid
HDAC10
Full length human recombit protein of HDAC10 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit