Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC2 Rabbit anti-Canine, Hamster, Human, Mouse, Non-human primate, Rat, Alexa Fluor® 488, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1861A488

Catalog No. PIPA1861A48


Add to cart

Description

Description

PA1861A488 detects histone deacetylase 2 (HDAC2) from canine, hamster, human, mouse, non-human primate, rat tissues and cells. The PA1861A488 immunogen is a synthetic peptide corresponding to residues C T(475) D T K G T K S E Q L S N P(488) of human HDAC2. This immunizing peptide (Cat. No. PEP-070) is available for use in neutralization and control experiments.

Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDA2 is likely to be involved in regulating chromatin structure during transcription. It has been implicated to associate with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA as a cofactor. This process is highly concerved from yeast to human.
Specifications

Specifications

HDAC2
Polyclonal
Alexa Fluor™ 488
HDAC2
Liquid
HDAC2
Synthetic peptide corresponding to residues C T(475) D T K G T K S E Q L S N P(488) of human HDAC2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Hamster, Human, Mouse, Non-human primate, Rat
Immunocytochemistry, Immunofluorescence
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Q92769, P70288
mRPD3, HD2, D10Wsu179e, YAF1, Yy1bp, RPD3
Rabbit
IgG
50 μL
4° C, store in dark, DO NOT FREEZE!
Primary
100764557, 15182, 3066, 475035, 84577
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit