Learn More
HDAC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 161521AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0756
Add to cart
Description
Description
This antibody is a rabbit polyclonal antibody raised against a peptide mapping within human HDAC2 and is specific to HDAC2. It will not cross react with other HDACs.
The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein shows sequence similarity to the E. coli RNA polymerase-binding protein HepA. Mutations in this gene are a cause of Schimke immunoosseous dysplasia (SIOD), an autosomal recessive disorder with the diagnostic features of spondyloepiphyseal dysplasia, renal dysfunction, and T-cell immunodeficiency.
Specifications
Specifications
HDAC2
Polyclonal
Unconjugated
HDAC2
HD2, HDAC2, histone deacetylase 2, RPD3, YAF1
Rabbit
Antigen Affinity Chromatography
RUO
15182, 3066, 84577
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70288, Q92769
Hdac2
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG