Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC3 Mouse anti-Human, Alexa Fluor 532, Clone: PCRP-HDAC3-2D4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314060AF532
This item is not returnable.
View return policy
Description
HDAC3 Monoclonal antibody specifically detects HDAC3 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
HDAC3 | |
Monoclonal | |
Alexa Fluor 532 | |
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 | |
Recombinant full-length human HDAC3 protein (Uniprot: O15379) | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics | |
8841 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-HDAC3-2D4 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title