Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC3 Rabbit anti-Cynomolgus Monkey, Human, Mouse, Rat, Clone: 8H2L16, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 702440

Encompass

Catalog No. 70-244-0


Add to cart

Description

Description

This antibody is predicted to react with Pig, Rat, Rabbit Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

HDAC3 Recombinant Monoclonal antibody specifically detects HDAC3 in Cynomolgus Monkey, Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

HDAC3
Western Blot
8H2L16
Unconjugated
HDAC3
AW537363; HD3; HDAC3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; histone deacetylase 3; histone deacetylase 3-like protein; RPD3; RPD3-2; SMAP45
Protein corresponding to human HDAC3
Protein A
RUO
Antibody
Monoclonal
Cynomolgus Monkey, Human, Mouse, Rat
Liquid
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O15379, O88895, Q6P6W3
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
102119873, 15183, 84578, 8841
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit