Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC3 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™

Rabbit Polyclonal Antibody
Supplier: Active Motif 40968

Description
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.Specifications
HDAC3 | |
Polyclonal | |
Unconjugated | |
HDAC3 | |
AW537363; HD3; Hdac3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; Histone deacetylase 3; histone deacetylase 3-like protein; RPD3; RPD3-2; SMAP45 | |
Rabbit | |
Protein G | |
RUO | |
8841 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Lot-Specific | |
PBS with 0.05% sodium azide; pH 7.5 | |
O15379 | |
HDAC3 | |
This HDAC3 antibody was raised against a synthetic peptide corresponding to amino acid residues 2-17 of human HDAC3. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction