Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC6 Mouse anti-Human, Monkey, Allophycocyanin/Cy7, Clone: OTI1B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271531APCCY7

Catalog No. N271531APC7


Add to cart

Description

Description

HDAC6 Monoclonal antibody specifically detects HDAC6 in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

HDAC6
OTI1B3
Flow Cytometry
Affinity Purified, Conjugated
HDAC6
Full length recombinant protein of human HDAC6 (NP_006035) produced in HEK293T cells.
Immunogen affinity purified
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Neuroscience
Primary
10013.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit