Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC6 Mouse anti-Human, Primate, Monkey, APC, Clone: OTI1B3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271531APC
Description
HDAC6 Monoclonal specifically detects HDAC6 in Human, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
HDAC6 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
HDAC6 | |
Full length recombinant protein of human HDAC6 (NP_006035) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Primate, Monkey | |
IgG1 |
Flow Cytometry | |
OTI1B3 | |
Flow Cytometry | |
EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
Mouse | |
Affinity Purified | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Neuroscience | |
10013.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title