Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC6, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A301342AM

 View more versions of this product

Catalog No. 50-156-0137


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
Specifications

Specifications

HDAC6
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q9UBN7, Q9Z2V5
FLJ16239, HD6, JM21
Rabbit
IgG
100 μl
4° C
Primary
10013, 15185
Western Blot
Unconjugated
HDAC6
liquid
HDAC6
Between 1165 and 1215
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit