Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC6 Rabbit anti-Human, Mouse, PE/Atto594, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP178981PEATT594
Description
HDAC6 Polyclonal antibody specifically detects HDAC6 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
HDAC6 | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Polyclonal | |
Human, Mouse |
Flow Cytometry | |
Flow Cytometry | |
Affinity Purified, Conjugated | |
HDAC6 | |
A synthetic peptide made to a C-terminal portion of the human HDAC6 protein (between residues 1175-1215). [UniProt Q9UBN7] | |
Immunogen affinity purified | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Neuroscience | |
Primary | |
10013.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title