Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$212.55 - $493.00
Specifications
Antigen | HDAC6 |
---|---|
Dilution | Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 20 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HDAC6 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Q9Z2V5 | |
10013 | |
Synthetic peptide mapping to the N-terminus of mouse HDAC-6. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
131 kDa |
Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 20 ug/mL | |
Polyclonal | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases | |
PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
HDAC6 | |
IgG | |
Affinity Purified | |
The antibody does not cross-react with other HDAC proteins including HDAC1, 2, 3, 4, 5, 7, and 8. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title