Learn More
HDAC6 Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI1D11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525326
Catalog No. PIMA525326
Add to cart
Description
Description
Histone deacetylase (HDAC) and histone acetyltransferase (HAT) are enzymes that regulate transcription by selectively deacetylating or acetylating the eta-amino groups of lysines located near the amino termini of core histone proteins. Eight members of HDAC family have been identified in the past several years. These HDAC family members are divided into two classes, I and II. Class I of the HDAC family comprises four members, HDAC-1, 2, 3, and 8, each of which contains a deacetylase domain exhibiting from 45 to 93% identity in amino acid sequence. Class II of the HDAC family comprises HDAC-4, 5, 6, and 7, the molecular weights of which are all about twofold larger than those of the class I members, and the deacetylase domains are present within the C-terminal regions, except that HDAC-6 contains two copies of the domain, one within each of the N-terminal and C-terminal regions. Human HDAC-1, 2 and 3 were expressed in various tissues, but the others (HDAC-4, 5, 6, and 7) showed tissue-specific expression patterns. These results suggested that each member of the HDAC family exhibits a different, individual substrate specificity and function in vivo.
Specifications
Specifications
HDAC6
Monoclonal
0.53 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UBN7
FLJ16239, HD6, JM21
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10013, 480907, 711363, 84581
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1D11
Unconjugated
HDAC6
Liquid
Hdac6
Full length human recombit protein of HDAC6 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat