Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC6 Mouse anti-Human, Non-human primate, Clone: OTI3E7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525359

Catalog No. PIMA525359


Add to cart

Description

Description

Histone deacetylase (HDAC) and histone acetyltransferase (HAT) are enzymes that regulate transcription by selectively deacetylating or acetylating the eta-amino groups of lysines located near the amino termini of core histone proteins. Eight members of HDAC family have been identified in the past several years. These HDAC family members are divided into two classes, I and II. Class I of the HDAC family comprises four members, HDAC-1, 2, 3, and 8, each of which contains a deacetylase domain exhibiting from 45 to 93% identity in amino acid sequence. Class II of the HDAC family comprises HDAC-4, 5, 6, and 7, the molecular weights of which are all about twofold larger than those of the class I members, and the deacetylase domains are present within the C-terminal regions, except that HDAC-6 contains two copies of the domain, one within each of the N-terminal and C-terminal regions. Human HDAC-1, 2 and 3 were expressed in various tissues, but the others (HDAC-4, 5, 6, and 7) showed tissue-specific expression patterns. These results suggested that each member of the HDAC family exhibits a different, individual substrate specificity and function in vivo.
Specifications

Specifications

HDAC6
Monoclonal
0.61 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UBN7
HD6, HDA6, Histone deacetylase 6, JM21
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10013, 711363
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3E7
Unconjugated
HDAC6
Liquid
HDAC6
Full length human recombit protein of HDAC6 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit