Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC9, Rabbit anti-Human,Mouse,Bovine,Canine,Primate, Alexa Fluor 750, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP203993AF750

Catalog No. NB004782


Add to cart

Description

Description

HDAC9 Polyclonal specifically detects HDAC9 in Human, Mouse, Bovine, Canine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

HDAC9
Alexa Fluor 750
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.5.1.98, HD7b, HD7HDAC, HD9, HDAC7, HDAC7BDKFZp779K1053, HDRP, histone deacetylase 4/5-related protein, Histone deacetylase 7B, histone deacetylase 9, Histone deacetylase-related protein, KIAA0744HDAC9B, MEF-2 interacting transcription repressor (MITR) protein, MEF2-interacting transcription repressor MITR, MITRHDAC9FL
Rabbit
IgG
0.1 mL
Cell Cycle and Replication, Epigenetics
Primary
Human, Mouse, Bovine, Canine, Primate
Western Blot
Western Blot
Affinity Purified, Conjugated
HDAC9
A portion of amino acids 100-150 of human HDAC-9 was used as the immunogen.
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit