Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDGFL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561067
Description
Immunogen sequence: ELREEEVADE EASQEWHAEA PGGGDRDS Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 43%.

Specifications
HDGFL1 | |
Polyclonal | |
Unconjugated | |
HDGFL1 | |
dJ309H15.1; HDGF (hepatoma-derived growth factor) like; HDGFL1; hepatoma derived growth factor-like 1; hepatoma-derived growth factor-like protein 1; PWWP domain containing 1; PWWP domain-containing protein 1; PWWP1; testicular tissue protein Li 84 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
154150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TGJ6 | |
HDGFL1 | |
Recombinant protein corresponding to Human HDGFL1. Recombinant protein control fragment (Product #RP-99853). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction