Learn More
HDHD1 Mouse anti-Canine, Human, Mouse, Non-human primate, Rat, Clone: OTI7A2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525456
Catalog No. PIMA525456
Add to cart
Description
Description
This gene encodes a member of the haloacid dehalogenase-like (HAD) hydrolase superfamily. The encoded protein has no known biological function. This gene has a pseudogene on chromosome 1. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Specifications
HDHD1
Monoclonal
1.2 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D5U5, Q08623
DXF68S1E, FAM16AX, GS1, HDHD1A, 5′-PsiMPase, family with sequence similarity 16, member A, X-linked, haloacid dehalogenase-like hydrolase domain containing 1A, haloacid dehalogenase-like hydrolase domain-containing protein 1, haloacid dehalogenase-like hydrolase domain-containing protein 1A
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
291585, 610678, 67365, 707930, 8226
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI7A2
Unconjugated
PUDP
Liquid
PUDP
Full length human recombit protein of HDHD1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Non-human primate, Rat