Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDHD2 Monoclonal Antibody (OTI1A4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525539

Catalog No. PIMA525539


Only null left
Add to Cart

Description

Description

HDHD2 Monoclonal Antibody for Western Blot

HDHD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDHD2
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H0R4
HDHD2
Full length human recombinant protein of HDHD2 produced in HEK293T cell.
100 μL
Primary
Human, Canine, Monkey
Antibody
IgG1
Western Blot
OTI1A4
Unconjugated
HDHD2
0610039H12Rik; 3110052N05Rik; epididymis secretory protein Li 301; haloacid dehalogenase like hydrolase domain containing 2; haloacid dehalogenase-like hydrolase domain containing 2; haloacid dehalogenase-like hydrolase domain-containing protein 2; hdhd2; HEL-S-301; hypothetical protein LOC505403; hypothetical protein LOC541346; Ier3ip1; ier3ip1 {ECO:0000250; immediate early response 3-interacting protein 1; LRRG00122; RGD1308579; UniProtKB:Q9Y5U9}; zgc:110332
Mouse
Affinity Chromatography
RUO
480146, 84064
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.