Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HDJ2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581929
Description
Immunogen sequence: DQKDHAVFTR RGEDLFMCMD IQLVEALCGF QKPISTLDNR TIVITSHPGQ IVKHGDIKCV LNEGMPIYRR PYEKGRLIIE FKVNFPENGF LSPDKLSLLE KLLPERKEVE ETDEMDQVEL VDFDPNQERR RHYNGE.
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1. 6 kb and a protein product with a molecular weight of about 35 kDa.
Specifications
HDJ2 | |
Polyclonal | |
Unconjugated | |
DNAJA1 | |
DJ-2; DjA1; DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ heat shock protein family (Hsp40) member A1; dnaJ homolog subfamily A member 1; DnaJ protein homolog 2; Dnaj2; DNAJA1; dnaJ-like protein 1; DnaJ-like protein 2; DnaJ-like subfamily A member 1; h HSJ-2; HDJ2; hDj-2; Heat shock 40 kDa protein 4; heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ2; HSJ-2; HSPF4; human DnaJ protein 2; I79_019991; LOC100346508; LOW QUALITY PROTEIN: dnaJ homolog subfamily A member 1; NEDD7; neural precursor cell expressed, developmentally down-regulated 7; neural precursor cell expressed, developmentally down-regulated gene 7; Rdj1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15502, 3301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P31689, P63037 | |
DNAJA1 | |
Recombinant protein corresponding to Human DNAJA1. Recombinant protein control fragment (Product #RP-95148). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction