Learn More
Athens Research And Technology IncSupplier Diversity Partner HDL LIPOPROTEINS CELL CULTURE DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 1216080412TC1MG
Lipoproteins, High Density, HP, Cell Culture: 1 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.1% EDTA; MW 175,000-360,000; Endotoxin Level: <=10 EU/mg. Native; Prepared & purified from fresh, non-frozen human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-859