Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ HDPE, Amber Carboy

Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
20 L
40 L

For Use With: Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications; Closure Material: Polypropylene; Color: Amber

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Material, volume, and metric graduation marks imprinted on carboys
Capacity (English) 10.5 gal.
Closure Material Polypropylene
Closure Size 120 mm
Capacity (Metric) 40 L
Dimensions (L x W x H) 434 x 249 x 772 mm
Material HDPE
Type Amber Carboy
For Use With (Application) Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications
Height (English) 30.4 in.
Length (English) 17.1 in.
Shape Rectangular
Width (English) 9.8 in.
Color Amber
Show More Show Less