Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDS 029, Tocris Bioscience™

Click to view available options
Quantity:
10mg
1mg
This item is not returnable. View return policy
881001-19-0
C17H11ClFN5O
355.757
DLPSDPPZXRJQOY-UHFFFAOYSA-N
egfr/erbb-2/erbb-4 inhibitor,unii-zn0x0q76f1,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl 2-butynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl but-2-ynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl-2-butynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl but-2-ynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl-2-butynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl but-2-ynamide,d05fft,tki-39
11566580
N-[4-(3-chloro-4-fluoroanilino)pyrido[3,4-d]pyrimidin-6-yl]but-2-ynamide
CC#CC(=O)NC1=NC=C2C(=C1)C(=NC=N2)NC3=CC(=C(C=C3)F)Cl
This item is not returnable. View return policy
881001-19-0
C17H11ClFN5O
355.757
DLPSDPPZXRJQOY-UHFFFAOYSA-N
egfr/erbb-2/erbb-4 inhibitor,unii-zn0x0q76f1,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl 2-butynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl but-2-ynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl-2-butynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl but-2-ynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl-2-butynamide,n-4-3-chloro-4-fluorophenyl amino pyrido 3,4-d pyrimidin-6-yl but-2-ynamide,d05fft,tki-39
11566580
N-[4-(3-chloro-4-fluoroanilino)pyrido[3,4-d]pyrimidin-6-yl]but-2-ynamide
CC#CC(=O)NC1=NC=C2C(=C1)C(=NC=N2)NC3=CC(=C(C=C3)F)Cl

Potent inhibitor of the ErbB receptor family

Potent inhibitor of the ErbB receptor family (IC50 values are 0.3, 0.5 and 1.1 nM for ErbB1 (EGFR), ErbB4 and ErbB2 respectively). Inhibits EGF-induced erbB1 autophosphorylation in NIH3T3 cells and heregulin-stimulated ErbB autophosphorylation in MDA-MB-453 human breast carcinoma cells (IC50 values are 2.5 and 24 nM respectively).
Quantity 10mg
Formula Weight 355.76
Percent Purity >97%
Chemical Name or Material HDS 029

Recommended Storage : Store at -20°C