HDS 029, Tocris

Potent inhibitor of the ErbB receptor family

Manufacturer: R&D Systems 2646/1

Quantity 1mg

 View more versions of this product

Catalog No. 26-461

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >97%
    Molecular Formula C17H11ClFN5O
    Formula Weight 355.76
    Synonym N-(4-[(-3-Chloro-4-fluorophenyl)amino]pyrido[3, 4-d]pyrimidin-6-yl-2-butynamide
    Chemical Name or Material HDS 029

    Specifications

    Formula Weight 355.76
    Quantity 1mg
    Assay Percent Range >97%
    Molecular Formula C17H11ClFN5O
    CAS 881001-19-0
    Synonym N-(4-[(-3-Chloro-4-fluorophenyl)amino]pyrido[3, 4-d]pyrimidin-6-yl-2-butynamide
    Recommended Storage Store at -20°C
    Chemical Name or Material HDS 029

    Store at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit