Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HE4/WFDC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | HE4/WFDC2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3ug/ml, Immunohistochemistry-Paraffin 5-7ug/ml∼ |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227299SS
![]() |
Novus Biologicals
NBP227299SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227299
![]() |
Novus Biologicals
NBP227299 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
HE4/WFDC2 Polyclonal specifically detects HE4/WFDC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HE4/WFDC2 | |
Western Blot 1-3ug/ml, Immunohistochemistry-Paraffin 5-7ug/ml∼ | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Q14508 | |
10406 | |
A portion of amino acids 1-50 of (human) HE4 was used as the immunogen for the antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5epididymal protein 4 | |
WFDC2 | |
IgG | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title