Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HE4/WFDC2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP149888
Description
HE4/WFDC2 Polyclonal specifically detects HE4/WFDC2 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
HE4/WFDC2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5epididymal protein 4 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody recognizes and epitope within the internal region.This antibody is specific to the internal region. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.2 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_006094.3 | |
WFDC2 | |
Peptide with sequence GAEKTGVCPELQADQ corresponding to internal region according to NP_006094.3. | |
0.1 mg | |
Cancer | |
10406 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction