Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HE4/WFDC2 Antibody Pair [HRP]
SDP

Monoclonal Antibody Pair

$832.00

Specifications

Assay Range 15.625 pg/mL
Assay Sensitivity 15.625-1000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB009847
SDP
View Documents
Novus Biologicals™
NBP27940415PLATES
15 Plates
Each of 1 for $832.00
Add to Cart
 
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human HE4/WFDC2.
Specifications

Specifications

15.625 pg/mL
Sandwich ELISA
Antibody Pairs
Human
dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5epididymal protein 4
WFDC2
  • Standard: Each vial contains 50 ng of recombinant Human HE4/WFDC2.
  • Capture Antibody: 1 mg/mL of mouse anti-Human HE4/WFDC2 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human HE4/WFDC2 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Cancer
    HE4/WFDC2
    15.625-1000 pg/mL
    HRP
    This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human HE4/WFDC2.
    Sandwich ELISA
    10406
    Rabbit/Mouse
    RUO
    Storage is content dependent.
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.05 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended.
    SDS
    Documents

    Documents

    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit