Learn More
WASP Project Head for LDM Clay Extruder
SCI_ED
WASP LDM Extruder head. A replacement for an existing LDM Clay System
Supplier:  WASP Project PS7021
Catalog No. S81221
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
LDM WASP Extruder is a ceramic extruder, a professional instrument for the deposition of dense fluid material as clay mixture and experimental materials. It is equipped with a screw system capable of regulating the output flow of the material, allowing a rapid flow interruction and a good retraction control.
Specifications
Specifications
STEM Project