Learn More
Thermo Scientific™ Heat-Sealable Polyethylene Bags
Protect specimens for mailing with Thermo Scientific™ Heat-Sealable Polyethylene Bags.
Manufacturer:  Thermo Scientific™ 840
 View more versions of this product
Catalog No. 84-0
  Check Availability
Add to cart
Specifications
Description
Protect specimens for mailing with Thermo Scientific™ Heat-Sealable Polyethylene Bags.
  • Heavy-duty polyethylene construction
  • Heat-sealable to ensure leak-resistant storage
Ordering Information:
  • Sold in packs of 100
Description & Specifications
Specifications
Polyethylene
7.6cm
12.7cm
3 in.
5 in.