Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Heat Shock Protein 60 (HSP60) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and electron microscopy experiments.

Specifications

Antigen Heat Shock Protein 60 (HSP60) Ab-1
Clone LK1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS120R1
View Documents
Thermo Scientific™
MS120R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and electron microscopy experiments with Thermo Scientific™ Heat Shock Protein 60 (HSP60) Ab-1, Mouse Monoclonal Antibody.

A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of heat shock proteins (hsp's) or stress response proteins (srp's). hsp60 is a potential antigen in a number of autoimmune diseases including human arthritis.

Host Species: Mouse

Clone: LK1

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Hamster and Chicken. Does not react with bacteria, helminths and spinach. Others not known.

Epitope: aa 383-447

Immunogen: Recombinant HSP60 protein

Molecular Weight: 60 kDa

Positive Control: BT474 or MAD109 cells, Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Heat Shock Protein 60 (HSP60) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Stress Response
Recombinant HSP60 protein
LK1
Monoclonal
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit