Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Heat Shock Protein 70 (HSP70) Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

$175.10 - $1008.00

Specifications

Antigen Heat Shock Protein 70 (HSP70) Ab-2
Clone W27
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 9
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS482B
View Documents
Thermo Scientific™
MS482B
Biotinylated Ab with BSA and Azide 1mL Each for $1,008.00
Add to cart
 
MS482B0
View Documents
Thermo Scientific™
MS482B0
Biotinylated Ab with BSA and Azide 100μL Each for $232.00
Add to cart
 
MS482B1
View Documents
Thermo Scientific™
MS482B1
Biotinylated Ab with BSA and Azide 500μL Each for $635.50
Add to cart
 
MS482P
View Documents
Thermo Scientific™
MS482P
Purified Ab with BSA and Azide 1mL Each for $807.00
Add to cart
 
MS482P0
View Documents
Thermo Scientific™
MS482P0
Purified Ab with BSA and Azide 100μL Each for $175.10
Add to cart
 
MS482P1
View Documents
Thermo Scientific™
MS482P1
Purified Ab with BSA and Azide 500μL Each for $486.00
Add to cart
 
MS482P1ABX
View Documents
Thermo Scientific™
MS482P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $537.00
Add to cart
 
MS482PABX
View Documents
Thermo Scientific™
MS482PABX
Ab without BSA/Azide (Frozen) 200μL Each for $887.50
Add to cart
 
MS482R7
View Documents
Thermo Scientific™
MS482R7
Ready-to-use for IHC (UVLP) 7mL Each for $419.00
Add to cart
 
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ Heat Shock Protein 70 (HSP70) Ab-2, Mouse Monoclonal Antibody.

Heat shock proteins (HSP's) or stress response proteins (SRP's) are synthesized in variety of environmental and pathophysiological stressful conditions. Many HSP's are involved in processes such as protein denaturation-renaturation, folding-unfolding, transport-translocation, activation-inactivation, and secretion. HSP70 is found to be associated with steroid receptors, actin, p53, polyoma T antigen, nucleotides, and other unknown proteins. Also, HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs, and other damaging conditions.

Host Species: Mouse

Clone: W27

Isotype: IgG2a

Species Reactivity: Human, Monkey and Cow. Others not known.

Epitope: Not determined

Immunogen: HSP70 protein from HeLa cells

Molecular Weight: 73 kDa (constitutive), 72 kDa (inducible)

Positive Control: BT474 or HeLa cells, Breast CA

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Heat Shock Protein 70 (HSP70) Ab-2
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Stress Response
HSP70 protein from HeLa cells
W27
Monoclonal
RUO
Primary
IgG2a
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit