Learn More
Heathrow Scientific™ Grab Rack™
Dump the contents, not your tubes. Fisherbrand™ Grab Rack™ uses silicon grips to securely hold fifty 5 to 10mL tubes in place, so you can empty all of the tubes at once for greater efficiency. Insert or remove upright tubes into the dual-level rack using gentle pressure.
Manufacturer:  Heathrow Scientific™ HS24371A
Catalog No. 22-267-053
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Not for cleaning in autoclave
Description
Description
  • Rack features silicon well grips that keep tubes securely in place
  • Double-layer ABS rack design holds tubes upright
  • Tubes do not fall out when rack is turned over, so you can discard all the contents at once
  • Tubes insert and extract with a gentle push or pull
  • Features a 5 x 10 well array; no assembly required
Specifications
Specifications
White
5 to 10mL tubes
ABS Plastic
259 x 127 x 49 mm
259 x 127 x 49mm
Silicon Grip
50 Wells