Learn More
Fisher Scientific™ Heating Thermal Mixer II PROMO
For precise sample preparation when working with DNA, RNA, plasmids, cultures and enzymatic reactions
 View more versions of this product
Catalog No. 11-700-176
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Thermal Mixer
11 in.
7.5 in.
19.05cm
7 in.
Two years
10° to 30°C
0 to 99 hours: 59 minutes: 59 seconds/hold
UL, CE, CSA approved
27.94cm
Conical Tube Blocks (15-600-333 and 15-600-332)
250-3000rpm, in increments of 50rpm
17.78cm
12VDC via 115/230VAC adapter
15°C below ambient to 100°C