Learn More
Fisherbrand™ Heating Thermal Mixer II PROMO
For precise sample preparation when working with DNA, RNA, plasmids, cultures and enzymatic reactions.
 View more versions of this product
Catalog No. 11-700-185
Add to cart
Includes: Includes: Block for 96 Well Plate and Block for 1.5mL Tubes (15-600-341 and 15-600-333)
Description
Description
  • Large color touch screen
  • Interchangeable blocks
  • Auto-recognition of blocks
  • Mixing and temperature control for small samples
  • For laboratory use
  • Designed by scientists for scientists
  • Easy to use and program
Intuitive menu navigation
  • Home display indicates step number and program name
  • Unit operating status is indicated by bright red text
  • “Stop, Play, Pause, Skip and Quick Mix” icons are easy to use
  • Instantly change parameters for one step
  • Turn off or on parameters as needed during process
  • Minimum and maximum parameters are automatically determined by block type
  • View and modify all steps on program screen
  • Automatically repeat one sequence as many times as needed
  • Use as many as steps as needed, up to five available
  • Easily select, launch, change, and save programs
  • Programs can have long text names for easy process recall
  • Turn on parameter lock to protect important processes
  • Stores up to 25 programs
Certifications
Power Pac is UL-listed, CSA-approved and CE-marked.
Warranty and Services
Two years
Specifications
Specifications
Thermal Mixer
11 in.
7.5 in.
19.05cm
7 in.
Two years
10° to 30°C
0 to 99 hours: 59 minutes: 59 seconds/hold
UL, CE, CSA approved
27.94cm
Conical Tube Block (15-600-333) and PCR Plate Block (15-600-341)
250-3000rpm, in increments of 50rpm
17.78cm
12VDC via 115/230VAC adapter
15°C below ambient to 100°C