Learn More
Brady™ Heavy Duty Blank Fluorescent Tags
Identify facility equipment to your exact specifications. The Brady™ Heavy Duty Blank Fluorescent Tags are durable one-sided tags with a write-on area that accepts most marking pens.
Supplier: Brady™ 102054
Description
- Heavy duty paper tags are made of a durable cardstock material
- They provide an economical and convenient way to ID equipment or areas of your facility
- Paper grommet prevents tearing from the point of attachment
- Tag includes a write-on area that accepts most marking pens
- Tags are appropriate for indoor use
Specifications
1000/Pk. | |
Cardstock | |
2.875 x 5.75 in. |
Orange | |
Rectangle | |
Not laminated |