Learn More
Brady™ Heavy Duty Blank Fluorescent Tags
Identify facility equipment to your exact specifications. The Brady™ Heavy Duty Blank Fluorescent Tags are durable one-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 102066
 View more versions of this product
Catalog No. 19-112-978
Add to cart
Description
Description
  • Heavy duty paper tags are made of a durable cardstock material
  • They provide an economical and convenient way to ID equipment or areas of your facility
  • Paper grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor use
Specifications
Specifications
Green
Not laminated
Rectangle
1.875 in. x 3.75 in.
1000/Pack
Cardstock