Learn More
Brady™ Heavy Duty Blank Tags
Identify facility equipment to your exact specifications. The Brady™ Heavy Duty Blank Tags are durable one-sided tags with a write-on area that accepts most marking pens.
Supplier: Brady™ 102101
Description
- Heavy duty paper tags are made of a durable cardstock material
- They provide an economical and convenient way to ID equipment or areas of your facility
- Paper grommet prevents tearing from the point of attachment
- Tag includes a write-on area that accepts most marking pens
- Tags are appropriate for indoor use
Specifications
1000/Pk. | |
Cardstock | |
2.625 x 5.25 in. |
Dark Blue | |
Rectangle | |
Not laminated |