Learn More
3M™ Heavy Duty Duct Tape
3M™ Heavy Duty Duct Tape is constructed of polyethylene film laminated to cloth with a rubber adhesive
Manufacturer:  3M™ 00051131069756
Catalog No. 19-072-083
Add to cart
Description
Description
  • Designed for heavy duty use in MRO/construction applications
  • Sticks immediately and stays on
  • Resists curling and tears off the roll cleanly for easy application
  • Unique construction that allows for permanent and temporary applications.
  • Strong water-resistant backing resists wear and light abrasion
  • Provides a tight, moisture-resistant seal in many containment situations
  • Conforms to irregular surfaces
  • Low tack roll edges stay cleaner
  • Save time and hassle of removing sticky or dried-on residue
Specifications
Specifications
9 mil
Industrial Tape
Communications, Transportation
Individually packaged
Rubber
Silver
48 mm x 54.8 m
Rubber
24/case