Learn More
Heavy-Duty Specimen Storage Bags
Keep plenty of these bags on hand for fast cleanup and storage after lab.
$22.74 - $52.25
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||
S6015
![]() |
Ranaco Corporation
03.0012.25 |
38 in. | 96.5cm | 38 x 12 in. (96.5 x 30.4cm) |
Pack of 25 for $52.25
|
|
S6016
![]() |
Ranaco Corporation
03.0032.25 |
20 in. | 50.8cm | 20 x 12 in. (50.8 x 30.4cm) |
Pack of 25 for $22.74
|
|
Description
- 38 x 12 in. (96.5 x 30.4cm) bags for storage of cats, rabbits, sharks, etc.
- 20 x 12 in. (50.8 x 30.4cm) bags for storage of fetal pigs, rats, etc.
- 12 x 12 in. (30.4 x 30.4cm) bags for storage of grass frogs, starfish, perch, etc.
Order Info
Come in packs of 25.
Specifications
12 in. | |
30.4cm |