Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heavy Magnesium Oxide, Heavy, Powder, FCC, 96-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1309-48-4
MgO
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
O=[Mg]
1309-48-4
MgO
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
O=[Mg]

M1168, 1309-48-4, MgO

Spectrum™ Heavy Magnesium Oxide, Heavy, Powder, FCC is a natural, GRAS ingredient used as an anti-caking agent, firming agent and for nutrition fortification. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 96 to 100.5%
Calcium Oxide 0.015
Insoluble in Acid 0.001
Lead (Pb) 4 mg/kg
Loss on Ignition 0.1
Packaging Poly Bottle
Quantity 500 g