Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Heavy Wall Cylindrical Pressure Vessels
Cylindrical vessels feature a protective coating with a working temperature ranging from -40°C to 80°C.
Supplier: Synthware™ P160004D
Description
- Protective coating has a working temperature ranging from -40°C to 80°C
- Coating may yellow over time, but will not affect the vessel's integrity
- Complete apparatus is supplied with a vessel, Teflon™ bushing, and an EP™ O-ring
- Always work behind a safety shield since there is a possibility that glass vessels can break under pressure
Specifications
#15 | |
70 mm | |
88 psi | |
Protective | |
46 mm |
1.6 oz. | |
Teflon™ Bushing, EP™ O-Ring | |
Cylindrical | |
48 mL | |
Glass |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction