Learn More
HECTD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206051AP
Catalog No. 50-173-2985
Add to cart
Description
Description
Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
Specifications
Specifications
HECTD1
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q69ZR2, Q9ULT8
E3 ligase for inhibin receptor, EULIR, HECT domain containing 1, HECTD1, KIAA1131
Rabbit
IgG
150 μL
-20°C
Primary
207304, 25831
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
HECTD1
Liquid
HECTD1
HECTD1 Fusion Protein Ag14581
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse