Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HECTD2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | HECTD2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155080
|
Novus Biologicals
NBP155080 |
100 μL |
Each of 1 for $436.00
|
|
Description
HECTD2 Polyclonal specifically detects HECTD2 in Human samples. It is validated for Western Blot.Specifications
HECTD2 | |
Polyclonal | |
Rabbit | |
Q5U5R9 | |
143279 | |
Synthetic peptides corresponding to HECTD2(HECT domain containing 2) The peptide sequence was selected from the middle region of HECTD2. Peptide sequence ALMLLRPEEVEILVCGSPDLDMHALQRSTQYDGYAKTDLTIKYFWDVVLG. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
EC 6.3.2.-, FLJ16050, FLJ37306, HECT domain containing 2, HECT domain-containing protein 2, probable E3 ubiquitin-protein ligase HECTD2 | |
HECTD2 | |
IgG | |
Affinity Purified | |
88 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title