Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HECTD3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548860

Catalog No. PA548860

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

E3 ubiquitin ligases accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Mediates ubiquitination of TRIOBP and its subsequent proteasomal degradation, thus faciliting cell cycle progression by regulating the turn-over of TRIOBP. Mediates also ubiquitination of STX8.
Specifications

Specifications

HECTD3
Polyclonal
PBS with 0.09% sodium azide
Q5T447
E3 ubiquitin-protein ligase HECTD3, 632-, HECT domain-containing protein 3, HECTD3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79654
Western Blot
Unconjugated
HECTD3
Liquid
HECTD3
KLH conjugated synthetic peptide between 740-769 amino acids from the C-terminal region of human HECTD3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit