Learn More
K1 Series Heelstop - Anti-slip Heel Traction Aid - Intrinsic
The K1 Series Heelstop is made with ice-penetrating tungsten carbide spikes, TR freeze-resistant rubber, and reflective hi-viz strips.
Supplier: K1 Series V5770170M
This item is not returnable.
View return policy
Description
- Built with the toughest materials
- Ice-penetrating tungsten carbide spikes cover the entire heel for maximum slip resistance
- Intrinsic design is certified to prevent sparking when working in areas where combustible materials may be present
- Proud to be the only fully certified ice-cleat brand in North America
- Freeze-resistant industrial quality TR rubber remains flexible even in the most extreme cold weather conditions
- Reflective strips provide added safety when working in low lit working environments
- Easy to attach and remove with convenient Velcro straps
- Unique design allows driving and climbing without interference
Specifications
Hook & Loop | |
Outdoor Surfaces, Ice Surfaces | |
Medium | |
Reusable | |
Yes |
Black | |
Vulcanized Rubber | |
Leather | |
Unisex |