Learn More
Sklar InstrumentsSupplier Diversity Partner TC Olsen-Hegar Needle Holder, Sklar, 4-3/4 in., Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218026
TC Olsen-Hegar Needle Holder, Sklar, 4-3/4 in., Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's Olsen-Hegar TC Needle Holder is a ratcheted, finger ring instrument that also has built in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with smooth tips and an overall length of 4-3/4 inches.
Catalog No. 50-143-7714