Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Olsen Hegar Needle Holder, Sklar, 6 in., Blunt, Straight, Cross-Serrated, Finger Rings, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21877
Econo Olsen Hegar Needle Holder, Sklar, 6 in., Blunt, Straight, Cross-Serrated, Finger Rings, Stainless Steel. The Econo Olsen-Hegar Needle Holder is a ratcheted, finger ring instrument that also has built in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with cross-serrated tips, and a central groove. It is 6 inches long and sold in boxes of 12.
Catalog No. 50-143-7833