Learn More
Sklar InstrumentsSupplier Diversity Partner Hegar Uterine Dilator, Sklar, 7 in., Straight, Single End 1mm, Sklar's Hegar Single-Ended Uterine Dilator is a manual, probe-style instrument used to dilate the cervix. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 904784
Hegar Uterine Dilator, Sklar, 7 in., Straight, Single End 1mm, Sklar's Hegar Single-Ended Uterine Dilator is a manual, probe-style instrument used to dilate the cervix. They are solid, nonflexible, and available in multiple diameters. These dilators are slightly curved and have smooth, rounded tips. This product is the 1mm dilator. The overall length is 7 inches.
Catalog No. 50-143-6719