Learn More
Sklar InstrumentsSupplier Diversity Partner Hegar Uterine Dilator, Sklar, Single End 3.5mm, Straight, 7 in., Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 904789
Hegar Uterine Dilator, Sklar, Single End 3.5mm, Straight, 7 in., Stainless Steel. Sklar's Hegar Single-Ended Uterine Dilator is a manual, probe-style instrument used to dilate the cervix. They are solid, nonflexible, and available in multiple diameters. These dilators are slightly curved and have smooth, rounded tips. This product is the 3.5mm dilator. The overall length is 7 inches.
Catalog No. 50-143-6724