Learn More
Heidolph™ Baffled Wide Neck Flasks for Radleys Tornado™ IS6 Overhead Stirring System and Carousel 6 Reaction Station™
Baffled wide neck flasks for use with Tornado™ Stirring System Carousel 6 Plus Reaction Station™
Manufacturer:  Heidolph™ 015862000
 View more versions of this product
Catalog No. 05-403-881
Add to cart
Description
Description
  • Wide neck flasks allow easier removal of viscous or solid samples and facilitate the use of the anchor and propeller style PTFE stirrer blade options (Tornado)
  • One or two side arms ideal for attaching dropping funnels or powder funnels.
  • Baffled flasks improve the turbulence within the flask by disrupting the creation of a central vortex.
Specifications
Specifications
8.45 oz.
Tornado™ IS6 Overhead Stirring System and Carousel 6 Reaction Station™
Flask
250 mL
B24/29 side arm and septa port