Learn More
Heidolph™ Duomax Rocking Orbit Platform Shaker Available on GSA/VA Contract for Federal Government customers only.
Platform lab shaker with rocking motion accepts load capacities of 5kg
Manufacturer:  Heidolph North America 036130280
 View more versions of this product
Catalog No. 13-878-459
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Duomax 1030
0 to 50°C
11.03 lb.
10°
7.2 in.
12.5 in.
37.5cm
17.63 lb.
115w
Acoustic
115V 50Hz
2 to 50rpm
115V
5kg
1 to 120 min.
14.75 in.
18.5cm
32cm
8kg
50Hz
14.75 x 12.5 x 7.2 in. (37.5 x 32 x 18.5cm)