Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heidolph™ Glassware - Baffled Flasks
Wide Neck Flasks with Baffles and Sidearms
$866.00 - $1830.00
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Includes | Price | Quantity & Availability | |||
13-900-157
|
Heidolph™
015861979 |
100 mL | B14/23 Side | 2xB14/23 Sidearm and Septa Port |
Each for $1,830.00
|
|
|||
13-900-156
|
Heidolph™
015861978 |
100 mL | B14/23 Side | B14/23 Sidearm and Septa Port |
Each for $866.00
|
|
|||
13-900-158
|
Heidolph™
015861981 |
250 mL | B24/29 Side | B24/29 Sidearm and Septa Port |
Each for $1,370.00
|
|
|||
13-900-159
|
Heidolph™
015861982 |
250 mL | B24/29 Side | 2xB24/29 Sidearm and Septa Port |
Each for $1,142.00
|
|
|||
Specifications
Glass | |
Wide |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title